Ethyl 5-amino-4-iodo-1-(p-tolyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480052-1G
APOSOR480052-1GEA 912 EUR
APOSOR480052-1G
Ethyl 5-amino-4-iodo-1-(p-tolyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-iodo-1-(p-tolyl)pyrazole-3-carboxylate
Formel: C₁₃H₁₄IN₃O₂
Molecular Weight: 371,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135221
CAS-Nummer: 1427010-79-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR