Ethyl 5-amino-4-chloro-1-(4-chlorophenyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480053-1G
APOSOR480053-1GEA
797
EUR
APOSOR480053-1G
Ethyl 5-amino-4-chloro-1-(4-chlorophenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(4-chlorophenyl)pyrazole-3-carboxylate
Formel:
C₁₂H₁₁Cl₂N₃O₂ Molecular Weight: 300,141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135222 CAS-Nummer: 1427012-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...