Ethyl 5-amino-1-(4-chlorophenyl)-4-iodo-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480055-1G
APOSOR480055-1GEA 912 EUR
APOSOR480055-1G
Ethyl 5-amino-1-(4-chlorophenyl)-4-iodo-pyrazole-3-carboxylate
Ethyl 5-amino-1-(4-chlorophenyl)-4-iodo-pyrazole-3-carboxylate
Formel: C₁₂H₁₁ClIN₃O₂
Molecular Weight: 391.592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135224
CAS-Nummer: 1427024-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR