Ethyl 5-amino-1-(3-bromophenyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480056-1G
APOSOR480056-1GEA
1090
EUR
APOSOR480056-1G
Ethyl 5-amino-1-(3-bromophenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(3-bromophenyl)pyrazole-3-carboxylate
Formel:
C₁₂H₁₂BrN₃O₂ Molecular Weight: 310,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22107823 CAS-Nummer: 1427020-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...