Ethyl 5-amino-1-(2-methylsulphonylphenyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480057-1G
APOSOR480057-1GEA
1770
EUR
APOSOR480057-1G
Ethyl 5-amino-1-(2-methylsulphonylphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-methylsulphonylphenyl)pyrazole-3-carboxylate
Formel:
C₁₃H₁₅N₃O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135147 CAS-Nummer: 1427021-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...