Ethyl 5-amino-1-(3-methylsulphonylphenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480058-1G
APOSOR480058-1GEA 1770 EUR
APOSOR480058-1G
Ethyl 5-amino-1-(3-methylsulphonylphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(3-methylsulphonylphenyl)pyrazole-3-carboxylate
Formel: C₁₃H₁₅N₃O₄S
Molecular Weight: 309,341 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135149
CAS-Nummer: 1367842-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR