Ethyl 5-amino-1-(4-methylsulphonylphenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480059-1G
APOSOR480059-1GEA 1770 EUR
APOSOR480059-1G
Ethyl 5-amino-1-(4-methylsulphonylphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(4-methylsulphonylphenyl)pyrazole-3-carboxylate
Formel: C₁₃H₁₅N₃O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD23135150
CAS-Nummer: 1427022-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR