Ethyl 5-amino-1-(4-sulphamoylphenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480060-1G
APOSOR480060-1GEA 1320 EUR
APOSOR480060-1G
Ethyl 5-amino-1-(4-sulphamoylphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(4-sulphamoylphenyl)pyrazole-3-carboxylate
Formel: C₁₂H₁₄N₄O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD23135151
CAS-Nummer: 866838-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR