Ethyl 5-amino-1-(3-sulphamoylphenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480061-1G
APOSOR480061-1GEA 1320 EUR
APOSOR480061-1G
Ethyl 5-amino-1-(3-sulphamoylphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(3-sulphamoylphenyl)pyrazole-3-carboxylate
Formel: C₁₂H₁₄N₄O₄S
Molecular Weight: 310.329 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135152
CAS-Nummer: 1427011-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR