Ethyl 5-amino-1-(2-sulphamoylphenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480062-1G
APOSOR480062-1GEA 1320 EUR
APOSOR480062-1G
Ethyl 5-amino-1-(2-sulphamoylphenyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-sulphamoylphenyl)pyrazole-3-carboxylate
Formel: C₁₂H₁₄N₄O₄S
Molecular Weight: 310,329 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135153
CAS-Nummer: 1427011-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR