Ethyl 5-amino-4-chloro-1-(2-pyridyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480063-1G
APOSOR480063-1GEA 1200 EUR
APOSOR480063-1G
Ethyl 5-amino-4-chloro-1-(2-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(2-pyridyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₁ClN₄O₂
Molecular Weight: 266,684 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135155
CAS-Nummer: 1427021-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR