Ethyl 5-amino-4-bromo-1-(2-pyridyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480064-1G
APOSOR480064-1GEA 1280 EUR
APOSOR480064-1G
Ethyl 5-amino-4-bromo-1-(2-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(2-pyridyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₁BrN₄O₂
Molecular Weight: 311,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135156
CAS-Nummer: 1427021-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR