Ethyl 5-amino-1-(3-pyridyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480065-1G
APOSOR480065-1GEA 1030 EUR
APOSOR480065-1G
Ethyl 5-amino-1-(3-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(3-pyridyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₂N₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23135157
CAS-Nummer: 1427023-96-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR