Ethyl 5-amino-4-chloro-1-(3-pyridyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480066-1G
APOSOR480066-1GEA 1280 EUR
APOSOR480066-1G
Ethyl 5-amino-4-chloro-1-(3-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(3-pyridyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₁ClN₄O₂
Molecular Weight: 266,684 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135158
CAS-Nummer: 1427022-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR