Ethyl 5-amino-4-bromo-1-(3-pyridyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480067-1G
APOSOR480067-1GEA
1340
EUR
APOSOR480067-1G
Ethyl 5-amino-4-bromo-1-(3-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(3-pyridyl)pyrazole-3-carboxylate
Formel:
C₁₁H₁₁BrN₄O₂ Molecular Weight: 311.135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135160 CAS-Nummer: 1427013-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...