Ethyl 5-amino-1-(4-pyridyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480068-1G
APOSOR480068-1GEA 1030 EUR
APOSOR480068-1G
Ethyl 5-amino-1-(4-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(4-pyridyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₂N₄O₂
Molecular Weight: 232,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135161
CAS-Nummer: 1427013-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR