Ethyl 5-amino-4-chloro-1-(4-pyridyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480069-1G
APOSOR480069-1GEA 1200 EUR
APOSOR480069-1G
Ethyl 5-amino-4-chloro-1-(4-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-chloro-1-(4-pyridyl)pyrazole-3-carboxylate
Formel: C₁₁H₁₁ClN₄O₂
Molecular Weight: 266.684 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135162
CAS-Nummer: 1427021-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR