Ethyl 5-amino-4-bromo-1-(4-pyridyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480070-1G
APOSOR480070-1GEA
1200
EUR
APOSOR480070-1G
Ethyl 5-amino-4-bromo-1-(4-pyridyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(4-pyridyl)pyrazole-3-carboxylate
Formel:
C₁₁H₁₁BrN₄O₂ Molecular Weight: 311,135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135163 CAS-Nummer: 1427013-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...