Ethyl 5-amino-1-pyrimidin-4-yl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480071-1G
APOSOR480071-1GEA 1140 EUR
APOSOR480071-1G
Ethyl 5-amino-1-pyrimidin-4-yl-pyrazole-3-carboxylate
Ethyl 5-amino-1-pyrimidin-4-yl-pyrazole-3-carboxylate
Formel: C₁₀H₁₁N₅O₂
Molecular Weight: 233,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135164
CAS-Nummer: 1427019-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR