Ethyl 5-amino-1-(1-naphthyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480073-1G
APOSOR480073-1GEA 684 EUR
APOSOR480073-1G
Ethyl 5-amino-1-(1-naphthyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(1-naphthyl)pyrazole-3-carboxylate
Formel: C₁₆H₁₅N₃O₂
Molecular Weight: 281,309 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07388442
CAS-Nummer: 1427011-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR