Ethyl 5-amino-1-(2-naphthyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480074-1G
APOSOR480074-1GEA
797
EUR
APOSOR480074-1G
Ethyl 5-amino-1-(2-naphthyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(2-naphthyl)pyrazole-3-carboxylate
Formel:
C₁₆H₁₅N₃O₂ Molecular Weight: 281,309 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07388441 CAS-Nummer: 1427024-05-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...