Ethyl 5-amino-1-pyrazin-2-yl-pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR480075-1G
APOSOR480075-1GEA 1030 EUR
APOSOR480075-1G
Ethyl 5-amino-1-pyrazin-2-yl-pyrazole-3-carboxylate
Ethyl 5-amino-1-pyrazin-2-yl-pyrazole-3-carboxylate
Formel: C₁₀H₁₁N₅O₂
Molecular Weight: 233,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135165
CAS-Nummer: 1427021-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR