Ethyl 5-amino-1-(4-quinolyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480076-1G
APOSOR480076-1GEA
1770
EUR
APOSOR480076-1G
Ethyl 5-amino-1-(4-quinolyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(4-quinolyl)pyrazole-3-carboxylate
Formel:
C₁₅H₁₄N₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135166 CAS-Nummer: 1427013-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...