Ethyl 5-amino-1-(3-quinolyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480077-1G
APOSOR480077-1GEA
1770
EUR
APOSOR480077-1G
Ethyl 5-amino-1-(3-quinolyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(3-quinolyl)pyrazole-3-carboxylate
Formel:
C₁₅H₁₄N₄O₂ Molecular Weight: 282.297 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135167 CAS-Nummer: 1427022-90-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...