Ethyl 5-amino-1-(5-quinolyl)pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480079-1G
APOSOR480079-1GEA
1140
EUR
APOSOR480079-1G
Ethyl 5-amino-1-(5-quinolyl)pyrazole-3-carboxylate
Ethyl 5-amino-1-(5-quinolyl)pyrazole-3-carboxylate
Formel:
C₁₅H₁₄N₄O₂ Molecular Weight: 282,297 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135169 CAS-Nummer: 1427014-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...