Ethyl 5-amino-1-quinazolin-4-yl-pyrazole-3-carboxylate
Lieferant: Apollo Scientific
OR480080-1G
APOSOR480080-1GEA
1030
EUR
APOSOR480080-1G
Ethyl 5-amino-1-quinazolin-4-yl-pyrazole-3-carboxylate
Ethyl 5-amino-1-quinazolin-4-yl-pyrazole-3-carboxylate
Formel:
C₁₄H₁₃N₅O₂ Molecular Weight: 283,285 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23135170 CAS-Nummer: 1427021-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...