1-(Pyridin-3-yl)cyclopropanamine dihydrochloride

Lieferant: Apollo Scientific

OR480082-1G
APOSOR480082-1GEA 1200 EUR
APOSOR480082-1G
1-(Pyridin-3-yl)cyclopropanamine dihydrochloride
1-(Pyridin-3-yl)cyclopropanamine dihydrochloride
Formel: C₈H₁₀N₂·2ClH
Molecular Weight: 207,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08752627
CAS-Nummer: 1187932-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR