1-(4-Chloro-2-pyridyl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

OR480086-1G
APOSOR480086-1GEA 2280 EUR
APOSOR480086-1G
1-(4-Chloro-2-pyridyl)cyclopropanamine hydrochloride
1-(4-Chloro-2-pyridyl)cyclopropanamine hydrochloride
Formel: C₈H₉ClN₂·ClH
Molecular Weight: 205.084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496524
CAS-Nummer: 1384265-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR