1-(3-Chloro-2-pyridyl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

OR480087-1G
APOSOR480087-1GEA 2280 EUR
APOSOR480087-1G
1-(3-Chloro-2-pyridyl)cyclopropanamine hydrochloride
1-(3-Chloro-2-pyridyl)cyclopropanamine hydrochloride
Formel: C₈H₉ClN₂·ClH
Molecular Weight: 205,084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496527
CAS-Nummer: 1060811-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR