1-(2-Chloro-3-pyridyl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

OR480089-1G
APOSOR480089-1GEA 2280 EUR
APOSOR480089-1G
1-(2-Chloro-3-pyridyl)cyclopropanamine hydrochloride
1-(2-Chloro-3-pyridyl)cyclopropanamine hydrochloride
Formel: C₈H₉ClN₂·ClH
Molecular Weight: 205,084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496533
CAS-Nummer: 2149589-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR