1-(5-Chloro-3-pyridyl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

OR480090-1G
APOSOR480090-1GEA 2280 EUR
APOSOR480090-1G
1-(5-Chloro-3-pyridyl)cyclopropanamine hydrochloride
1-(5-Chloro-3-pyridyl)cyclopropanamine hydrochloride
Formel: C₈H₉ClN₂·ClH
Molecular Weight: 205.084 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496536
CAS-Nummer: 1256787-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR