1-(3-Methylimidazol-4-yl)cyclopropanamine hydrochloride
Lieferant: Apollo Scientific
OR480097-1G
APOSOR480097-1GEA
3420
EUR
APOSOR480097-1G
1-(3-Methylimidazol-4-yl)cyclopropanamine hydrochloride
1-(3-Methylimidazol-4-yl)cyclopropanamine hydrochloride
Formel:
C₇H₁₁N₃·ClH Molecular Weight: 173,643 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD30496553 CAS-Nummer: 2149589-57-9 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...