1-(1-Methylimidazol-4-yl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

OR480098-1G
APOSOR480098-1GEA 3420 EUR
APOSOR480098-1G
1-(1-Methylimidazol-4-yl)cyclopropanamine hydrochloride
1-(1-Methylimidazol-4-yl)cyclopropanamine hydrochloride
Formel: C₇H₁₁N₃·ClH
Molecular Weight: 173,643 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496556
CAS-Nummer: 1159737-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR