1H-Indazole-4-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR48010-1G
OR48010-500MG
APOSOR48010-1GEA
343
EUR
APOSOR48010-1G
APOSOR48010-500MG
1H-Indazole-4-carbonitrile
1H-Indazole-4-carbonitrile
Formel:
C₈H₅N₃ Molecular Weight: 143.15 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09261250 CAS-Nummer: 861340-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...