1-Pyridazin-3-ylcyclopropanecarbonitrile

Lieferant: Apollo Scientific

OR480104-1G
APOSOR480104-1GEA 3420 EUR
APOSOR480104-1G
1-Pyridazin-3-ylcyclopropanecarbonitrile
1-Pyridazin-3-ylcyclopropanecarbonitrile
Formel: C₈H₇N₃
Molecular Weight: 145,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496583
CAS-Nummer: 1870107-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR