1-(1-Methylpyrrol-2-yl)cyclopropanecarbonitrile
Lieferant: Apollo Scientific
OR480106-1G
APOSOR480106-1GEA
3420
EUR
APOSOR480106-1G
1-(1-Methylpyrrol-2-yl)cyclopropanecarbonitrile
1-(1-Methylpyrrol-2-yl)cyclopropanecarbonitrile
Formel:
C₉H₁₀N₂ Molecular Weight: 146,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134916 CAS-Nummer: 1427020-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...