1-Oxazol-4-ylcyclopropanecarbonitrile

Lieferant: Apollo Scientific

OR480107-1G
APOSOR480107-1GEA 3420 EUR
APOSOR480107-1G
1-Oxazol-4-ylcyclopropanecarbonitrile
1-Oxazol-4-ylcyclopropanecarbonitrile
Formel: C₇H₆N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD30496588
CAS-Nummer: 2104221-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR