1-Thiazol-4-ylcyclopropanecarbonitrile

Lieferant: Apollo Scientific

OR480108-1G
APOSOR480108-1GEA 3420 EUR
APOSOR480108-1G
1-Thiazol-4-ylcyclopropanecarbonitrile
1-Thiazol-4-ylcyclopropanecarbonitrile
Formel: C₇H₆N₂S
Molecular Weight: 150,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496590
CAS-Nummer: 2109909-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR