1-(3-Chloro-2-pyridyl)cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

OR480109-1G
APOSOR480109-1GEA 1940 EUR
APOSOR480109-1G
1-(3-Chloro-2-pyridyl)cyclopropanecarboxylic acid
1-(3-Chloro-2-pyridyl)cyclopropanecarboxylic acid
Formel: C₉H₈ClNO₂
Molecular Weight: 197,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134934
CAS-Nummer: 1427020-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR