1-Pyridazin-4-ylcyclopropanecarboxylic acid

Lieferant: Apollo Scientific

OR480110-1G
APOSOR480110-1GEA 2740 EUR
APOSOR480110-1G
1-Pyridazin-4-ylcyclopropanecarboxylic acid
1-Pyridazin-4-ylcyclopropanecarboxylic acid
Formel: C₈H₈N₂O₂
Molecular Weight: 164,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496513
CAS-Nummer: 2090420-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR