1-Thiazol-4-ylcyclopropanecarboxylic acid
Lieferant: Apollo Scientific
OR480111-1G
APOSOR480111-1GEA
1660
EUR
APOSOR480111-1G
1-Thiazol-4-ylcyclopropanecarboxylic acid
1-Thiazol-4-ylcyclopropanecarboxylic acid
Formel:
C₇H₇NO₂S Molecular Weight: 169,201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496514 CAS-Nummer: 1542332-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...