1-Thiazol-5-ylcyclopropanecarboxylic acid

Lieferant: Apollo Scientific

OR480112-1G
APOSOR480112-1GEA 1660 EUR
APOSOR480112-1G
1-Thiazol-5-ylcyclopropanecarboxylic acid
1-Thiazol-5-ylcyclopropanecarboxylic acid
Formel: C₇H₇NO₂S
Molecular Weight: 169,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496515
CAS-Nummer: 1509873-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR