1-Oxazol-5-ylcyclopropanecarboxylic acid
Lieferant: Apollo Scientific
OR480113-1G
APOSOR480113-1GEA
2280
EUR
APOSOR480113-1G
1-Oxazol-5-ylcyclopropanecarboxylic acid
1-Oxazol-5-ylcyclopropanecarboxylic acid
Formel:
C₇H₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496517 CAS-Nummer: 1539719-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...