1-(4-Bromopyridin-3-yl)cyclopropane-1-carbonitrile

Lieferant: Apollo Scientific

OR480118-1G
APOSOR480118-1GEA 2280 EUR
APOSOR480118-1G
1-(4-Bromopyridin-3-yl)cyclopropane-1-carbonitrile
1-(4-Bromopyridin-3-yl)cyclopropane-1-carbonitrile
Formel: C₉H₇BrN₂
Molecular Weight: 223,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496531
CAS-Nummer: 1780805-39-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR