1-(3-Bromopyridin-4-yl)cyclopropane-1-carbonitrile
Lieferant: Apollo Scientific
OR480119-1G
APOSOR480119-1GEA
2740
EUR
APOSOR480119-1G
1-(3-Bromopyridin-4-yl)cyclopropane-1-carbonitrile
1-(3-Bromopyridin-4-yl)cyclopropane-1-carbonitrile
Formel:
C₉H₇BrN₂ Molecular Weight: 223,069 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496534 CAS-Nummer: 1780794-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...