1-(3-Bromopyridin-2-yl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific

OR480120-1G
APOSOR480120-1GEA 2740 EUR
APOSOR480120-1G
1-(3-Bromopyridin-2-yl)cyclopropane-1-carboxylic acid
1-(3-Bromopyridin-2-yl)cyclopropane-1-carboxylic acid
Formel: C₉H₈BrNO₂
Molecular Weight: 242,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496537
CAS-Nummer: 1881884-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR