1-(4-Bromopyridin-2-yl)cyclopropane-1-carboxylic acid
Lieferant: Apollo Scientific
OR480121-1G
APOSOR480121-1GEA
2740
EUR
APOSOR480121-1G
1-(4-Bromopyridin-2-yl)cyclopropane-1-carboxylic acid
1-(4-Bromopyridin-2-yl)cyclopropane-1-carboxylic acid
Formel:
C₉H₈BrNO₂ Molecular Weight: 242,069 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496539 CAS-Nummer: 2091004-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...