1-(4-Bromopyridin-3-yl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific

OR480122-1G
APOSOR480122-1GEA 2120 EUR
APOSOR480122-1G
1-(4-Bromopyridin-3-yl)cyclopropane-1-carboxylic acid
1-(4-Bromopyridin-3-yl)cyclopropane-1-carboxylic acid
Formel: C₉H₈BrNO₂
Molecular Weight: 242,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496542
CAS-Nummer: 1785054-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR