1-(3-Bromopyridin-4-yl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific

OR480123-1G
APOSOR480123-1GEA 2740 EUR
APOSOR480123-1G
1-(3-Bromopyridin-4-yl)cyclopropane-1-carboxylic acid
1-(3-Bromopyridin-4-yl)cyclopropane-1-carboxylic acid
Formel: C₉H₈BrNO₂
Molecular Weight: 242,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30076364
CAS-Nummer: 1780834-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR