1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
Lieferant: Apollo Scientific
OR480142-1G
APOSOR480142-1GEA
4540
EUR
APOSOR480142-1G
1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
1-(5-Bromonaphthalen-1-yl)cyclopropane-1-carboxylic acid
Formel:
C₁₄H₁₁BrO₂ Molecular Weight: 291,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496581 CAS-Nummer: 1781079-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...